APOH316CDD20 Apollo Scientific Ltd

Cinnamamide

  • MDL number MFCD00008033
  • Cas Number : 621-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-150
Purity 97
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=130109

Safety Information

signalword Warning
pcodes P264;P270
MSDS http://www.apolloscientific.co.uk/display_item.php?id=130109
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart