APOH316CDD28 Apollo Scientific Ltd

5-Bromo-3-chloro-2-hydrazinopyridine

  • MDL number MFCD10000107
  • Cas Number : 1289076-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=132516

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=132516
Availability details for
Quantity
  • Availability
  • Add to Cart