APOH316CDD36 Apollo Scientific Ltd

2-Hydrazino-3-iodopyridine

  • MDL number MFCD10000048
  • Cas Number : 54231-42-4
  • Molecular Weight : 235.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-hydrazino-3-iodopyridine
Inchi Code 1S/C5H6IN3/c6-4-2-1-3-8-5(4)9-7/h1-3H,7H2,(H,8,9)
Inchi Key WRRJCOZVATYAGG-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-hydrazino-3-iodopyridine

Safety Information

signalword Warning
pcodes P280;P301+P312;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/2-hydrazino-3-iodopyridine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart