APOH316CDD37 Apollo Scientific Ltd

3-Bromo-5-chloro-2-hydrazinopyridine

  • MDL number MFCD10000080
  • Cas Number : 1289113-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=132514

Safety Information

signalword Warning
pcodes P280;P301+P312;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=132514
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart