APOH316CDD3A Apollo Scientific Ltd

1-Bromo-2-chloro-3,5-difluorobenzene

  • MDL number MFCD00135441
  • Cas Number : 187929-82-4
  • Molecular Weight : 227.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-2-chloro-3,5-difluorobenzene
Inchi Code 1S/C6H2BrClF2/c7-4-1-3(9)2-5(10)6(4)8/h1-2H
Inchi Key NYLNBNAPLADVTL-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Store at 2-8
Physical Form Solid-Fused
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-bromo-2-chloro-35-difluorobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-bromo-2-chloro-35-difluorobenzene
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart