APOH316CDD4E Apollo Scientific Ltd

1,2-Dibromo-3-chloro-5-fluorobenzene

  • MDL number MFCD09878108
  • Cas Number : 1000577-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=125771

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=125771
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart