APOH316CDD50 Apollo Scientific Ltd

1-Bromo-3-(difluoromethyl)-5-fluorobenzene

  • MDL number MFCD09261145
  • Cas Number : 627526-90-3
  • Molecular Weight : 225.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-3-(difluoromethyl)-5-fluorobenzene
Inchi Code 1S/C7H4BrF3/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3,7H
Inchi Key OPTDWHOFDCKSSE-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-bromo-3-difluoromethyl-5-fluorobenzene

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/1-bromo-3-difluoromethyl-5-fluorobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart