APOH316CDD52 Apollo Scientific Ltd

4'-Chloro-3'-fluorostyrene

  • MDL number MFCD20384550
  • Cas Number : 1263414-46-5
  • Molecular Weight : 156.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-chloro-2-fluoro-4-vinylbenzene
Inchi Code 1S/C8H6ClF/c1-2-6-3-4-7(9)8(10)5-6/h2-5H,1H2
Inchi Key ZCDHKYZAFKYYIU-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-fluorostyrene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-fluorostyrene
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart