APOH316CDD55 Apollo Scientific Ltd

4'-Bromo-2,2,2,2',6'-pentafluoroacetophenone

  • MDL number MFCD12922633
  • Cas Number : 1208074-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(4-Bromo-2,6-difluorophenyl)-2,2,2-trifluoroethanone
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=128781

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=128781
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart