APOH316CDD57 Apollo Scientific Ltd

4-Bromo-2,6-difluorobenzenesulfonyl chloride

  • MDL number MFCD06409102
  • Cas Number : 874804-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=125330

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=125330
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart