APOH316CDD59 Apollo Scientific Ltd

3-Bromo-2-fluoro-5-methylbenzoic acid

  • MDL number MFCD00506954
  • Cas Number : 72518-16-2
  • Molecular Weight : 233.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-2-fluoro-5-methylbenzoic acid
Inchi Code 1S/C8H6BrFO2/c1-4-2-5(8(11)12)7(10)6(9)3-4/h2-3H,1H3,(H,11,12)
Inchi Key JFAQBRSYTNDGBL-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-fluoro-5-methylbenzoic-acid

Safety Information

signalword Warning
pcodes P280;P301+P312;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-fluoro-5-methylbenzoic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart