APOH316CDD5F Apollo Scientific Ltd

2-Bromo-3,5-difluorobenzyl alcohol

  • MDL number MFCD28737913
  • Cas Number : 1256081-83-0
  • Molecular Weight : 223.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-bromo-3,5-difluorophenyl)methanol
Inchi Code 1S/C7H5BrF2O/c8-7-4(3-11)1-5(9)2-6(7)10/h1-2,11H,3H2
Inchi Key BUHABCPTQANVTG-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-35-difluorobenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart