APOH316CDD63 Apollo Scientific Ltd

Methyl 2-fluoro-4-methyl-5-nitrobenzoate

  • MDL number MFCD23863743
  • Cas Number : 753924-48-0
  • Molecular Weight : 213.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-fluoro-4-methyl-5-nitrobenzoate
Inchi Code 1S/C9H8FNO4/c1-5-3-7(10)6(9(12)15-2)4-8(5)11(13)14/h3-4H,1-2H3
Inchi Key GBECACDDVVYBHV-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-fluoro-4-methyl-5-nitrobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-fluoro-4-methyl-5-nitrobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart