APOH316CDD65 Apollo Scientific Ltd

2-Amino-6-(difluoromethyl)pyridine

  • MDL number MFCD25476529
  • Cas Number : 1315611-68-7
  • Molecular Weight : 144.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-(difluoromethyl)pyridin-2-amine
Inchi Code 1S/C6H6F2N2/c7-6(8)4-2-1-3-5(9)10-4/h1-3,6H,(H2,9,10)
Inchi Key YAVWHAMGXOMEMK-UHFFFAOYSA-N

Description

Synonym 6-(Difluoromethyl)pyridin-2-amine
Purity 98%
Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-6-difluoromethylpyridine

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-6-difluoromethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart