APOH316CDD68 Apollo Scientific Ltd

2-Bromo-4-chloro-6-fluorobenzeneboronic acid

  • MDL number MFCD28976797
  • Molecular Weight : 253.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-bromo-4-chloro-6-fluorophenyl)boronic acid
Inchi Code 1S/C6H4BBrClFO2/c8-4-1-3(9)2-5(10)6(4)7(11)12/h1-2,11-12H
Inchi Key FLBOJJPUUNTDGU-UHFFFAOYSA-N

Description

Synonym (2-Bromo-4-chloro-6-fluorophenyl)boronic acid
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-chloro-6-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-chloro-6-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart