APOH316CDD69 Apollo Scientific Ltd

2-(3-Aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol

  • MDL number MFCD17015636
  • Cas Number : 2402-67-7
  • Molecular Weight : 259.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol
Inchi Code 1S/C9H7F6NO/c10-8(11,12)7(17,9(13,14)15)5-2-1-3-6(16)4-5/h1-4,17H,16H2
Inchi Key LLDYYDVBNYAQJM-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-3-aminophenyl-111333-hexafluoropropan-2-ol

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/2-3-aminophenyl-111333-hexafluoropropan-2-ol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart