APOH316CDD6C Apollo Scientific Ltd

2-Bromo-6-fluoro-4-(trifluoromethyl)phenol

  • MDL number MFCD28739065
  • Cas Number : 1610471-16-3
  • Molecular Weight : 259

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-fluoro-4-trifluoromethylphenol

Safety Information

signalword Danger
pcodes P260;P280;P301+P330+P331
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-fluoro-4-trifluoromethylphenol
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart