APOH316CDD6F Apollo Scientific Ltd

6-Fluoro-5-iodopyridin-2-ol

  • MDL number MFCD16611044
  • Cas Number : 884660-48-4
  • Molecular Weight : 238.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-fluoro-5-iodo-2-pyridinol
Inchi Code 1S/C5H3FINO/c6-5-3(7)1-2-4(9)8-5/h1-2H,(H,8,9)
Inchi Key OSOZRTIYHZBEGL-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-fluoro-5-iodopyridin-2-ol

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/6-fluoro-5-iodopyridin-2-ol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart