APOH316CDD73 Apollo Scientific Ltd

2-Bromo-6-fluoro-4-(trifluoromethyl)benzaldehyde

  • MDL number MFCD31665800
  • Cas Number : 2092468-34-1
  • Molecular Weight : 271.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-fluoro-4-trifluoromethylbenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-fluoro-4-trifluoromethylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart