APOH316CDD7F Apollo Scientific Ltd

Ethyl 1-(4-fluorophenyl)-1,2,4-triazole-3-carboxylate

  • MDL number MFCD29905001
  • Cas Number : 1613022-95-9
  • Molecular Weight : 235.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C11H10FN3O2/c1-2-17-11(16)10-13-7-15(14-10)9-5-3-8(12)4-6-9/h3-7H,2H2,1H3
Inchi Key SZAZXQCELZVSPX-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-1-4-fluorophenyl-124-triazole-3-carboxylate

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/ethyl-1-4-fluorophenyl-124-triazole-3-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart