APOH316CDD8B Apollo Scientific Ltd

2-(2,2-Dimethylpropanoylamino)-6-(trifluoromethyl)benzoic acid

  • MDL number MFCD00097847

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=125509

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=125509
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart