APOH316CDD8C Apollo Scientific Ltd

5-Bromo-2-fluorobenzyl fluoride

  • MDL number MFCD31664909

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=121912

Safety Information

signalword Danger
pcodes P301+P330+P331;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=121912
pictograms GHS05;GHS08
hstatements H314;H351
Availability details for
Quantity
  • Availability
  • Add to Cart