APOH316CDD91 Apollo Scientific Ltd

2-Fluoro-N-methoxy-N-methyl-4-(trifluoromethyl)benzamide

  • MDL number MFCD27946933
  • Cas Number : 198967-27-0
  • Molecular Weight : 251.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
Inchi Code 1S/C10H9F4NO2/c1-15(17-2)9(16)7-4-3-6(5-8(7)11)10(12,13)14/h3-5H,1-2H3
Inchi Key NOUPPELZOLDUCK-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-n-methoxy-n-methyl-4-trifluoromethylbenzamide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-n-methoxy-n-methyl-4-trifluoromethylbenzamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart