APOH316CDD92 Apollo Scientific Ltd

Methyl 2-bromo-6-chloro-4-fluorobenzoate

  • MDL number MFCD28733926
  • Cas Number : 1695489-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=130153

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=130153
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart