APOH316CDD96 Apollo Scientific Ltd

4,8-Dichloro-5-fluoroquinoline

  • MDL number MFCD28010387
  • Cas Number : 1065093-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/48-dichloro-5-fluoroquinoline

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/48-dichloro-5-fluoroquinoline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart