APOH316CDD9E Apollo Scientific Ltd

4-(3-Chloro-4-fluorophenyl)-1H-1,2,4-triazol-5(4H)-one

  • MDL number MFCD31742298
  • Cas Number : 1346235-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=128517

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=128517
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart