APOH316CDDA2 Apollo Scientific Ltd

6-Fluorobenzofuran

  • MDL number MFCD28654686
  • Cas Number : 24410-60-4
  • Molecular Weight : 136.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-fluorobenzofuran
Inchi Code 1S/C8H5FO/c9-7-2-1-6-3-4-10-8(6)5-7/h1-5H
Inchi Key MUNCFYFZUDHPAG-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-fluorobenzofuran

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-fluorobenzofuran
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart