APOH316CDDA6 Apollo Scientific Ltd

2-Methoxy-6-(trifluoromethyl)pyrimidin-4(3H)-one

  • MDL number MFCD16988116
  • Cas Number : 175354-56-0
  • Molecular Weight : 194.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methoxy-6-(trifluoromethyl)-4(3H)-pyrimidinone
Inchi Code 1S/C6H5F3N2O2/c1-13-5-10-3(6(7,8)9)2-4(12)11-5/h2H,1H3,(H,10,11,12)
Inchi Key ILOOCRYHYUMTKL-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methoxy-6-trifluoromethylpyrimidin-43h-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-methoxy-6-trifluoromethylpyrimidin-43h-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart