APOH316CDDAA Apollo Scientific Ltd

Ethyl 8-fluoroimidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD13195311
  • Cas Number : 1260843-88-6
  • Molecular Weight : 208.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 8-fluoroimidazo[1,2-a]pyridine-2-carboxylate
Inchi Code 1S/C10H9FN2O2/c1-2-15-10(14)8-6-13-5-3-4-7(11)9(13)12-8/h3-6H,2H2,1H3
Inchi Key CRHIHOAPJLWZFT-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-8-fluoroimidazo12-apyridine-2-carboxylate

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/ethyl-8-fluoroimidazo12-apyridine-2-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart