APOH316CDDAB Apollo Scientific Ltd

8-Fluoroimidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD09994934
  • Cas Number : 1020034-56-3
  • Molecular Weight : 180.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-fluoroimidazo[1,2-a]pyridine-2-carboxylic acid
Inchi Code 1S/C8H5FN2O2/c9-5-2-1-3-11-4-6(8(12)13)10-7(5)11/h1-4H,(H,12,13)
Inchi Key QRVQMNMTBXGPQA-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-fluoroimidazo12-apyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P304+P340;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/8-fluoroimidazo12-apyridine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart