APOH316CDDB6 Apollo Scientific Ltd

8-Chloro-7-fluoroquinolin-4(1H)-one

  • MDL number MFCD31745743
  • Cas Number : 1065092-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/8-chloro-7-fluoroquinolin-41h-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/8-chloro-7-fluoroquinolin-41h-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart