APOH316CDDBD Apollo Scientific Ltd

2-(Fluoromethyl)aniline, N-BOC protected

  • MDL number MFCD31664917

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=122079

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=122079
pictograms GHS05;GHS08
hstatements H314;H351
Availability details for
Quantity
  • Availability
  • Add to Cart