APOH316CDDC0 Apollo Scientific Ltd

8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one

  • MDL number MFCD12406875
  • Cas Number : 1428532-93-7
  • Molecular Weight : 237.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-chloro-6-trifluoromethyl-124triazolo43-apyridin-32h-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/8-chloro-6-trifluoromethyl-124triazolo43-apyridin-32h-one
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart