APOH316CDDCE Apollo Scientific Ltd

2,6-Dibromo-4-fluoro-1,3-benzothiazole

  • MDL number MFCD09748620
  • Cas Number : 1188226-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=125701

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=125701
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart