APOH316CDDD7 Apollo Scientific Ltd

Ethyl 2-(4-Chlorophenyl)-2,2-difluoroacetate

  • MDL number MFCD11007726
  • Cas Number : 130754-19-7
  • Molecular Weight : 234.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl (4-chlorophenyl)(difluoro)acetate
Inchi Code 1S/C10H9ClF2O2/c1-2-15-9(14)10(12,13)7-3-5-8(11)6-4-7/h3-6H,2H2,1H3
Inchi Key VGULHXRXMXNYHN-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-4-chlorophenyl-22-difluoroacetate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-4-chlorophenyl-22-difluoroacetate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart