APOH316CDDDF Apollo Scientific Ltd

5-Bromo-4-fluoro-1,3-benzothiazol-2-amine

  • MDL number MFCD23705700
  • Cas Number : 1427383-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=125705

Safety Information

signalword Warning
pcodes P280;P301+P312;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=125705
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart