APOH316CDDE4 Apollo Scientific Ltd

2-(2-Fluoroethoxy)-5-methylbenzoic acid

  • MDL number MFCD20661084
  • Cas Number : 1269797-08-1
  • Molecular Weight : 198.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-fluoroethoxy)-5-methylbenzoic acid
Inchi Code 1S/C10H11FO3/c1-7-2-3-9(14-5-4-11)8(6-7)10(12)13/h2-3,6H,4-5H2,1H3,(H,12,13)
Inchi Key HPJMZARXICLMRD-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-2-fluoroethoxy-5-methylbenzoic-acid

Safety Information

signalword Warning
pcodes P280;P302+P352
MSDS https://store.apolloscientific.co.uk/product/2-2-fluoroethoxy-5-methylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart