APOH316CDDFE Apollo Scientific Ltd

Ethyl 4-[(3-chloro-4-methoxybenzyl)amino]-2-(methylthio)pyrimidine-5-carboxylate

  • MDL number MFCD17012695
  • Cas Number : 330785-81-4
  • Molecular Weight : 367.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-[(3-chloro-4-methoxybenzyl)amino]-2-(methylsulfanyl)-5-pyrimidinecarboxylate
Inchi Code 1S/C16H18ClN3O3S/c1-4-23-15(21)11-9-19-16(24-3)20-14(11)18-8-10-5-6-13(22-2)12(17)7-10/h5-7,9H,4,8H2,1-3H3,(H,18,19,20)
Inchi Key WLOQDGSMJNYRRC-UHFFFAOYSA-N

Description

Synonym Ethyl 4-[(3-chloro-4-methoxybenzyl)amino]-2-(methylsulphanyl)pyrimidine-5-carboxylate, 4-[(3-Chloro-4-methoxybenzyl)amino]-5-(ethoxycarbonyl)-2-(methylthio)pyrimidine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-4-3-chloro-4-methoxybenzylamino-2-methylthiopyrimidine-5-carboxylate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-4-3-chloro-4-methoxybenzylamino-2-methylthiopyrimidine-5-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart