APOH316CDE0E Apollo Scientific Ltd

2-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)-2,2-difluoroacetohydrazide

  • MDL number MFCD29042511
  • Cas Number : 1823183-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131309

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=131309
Availability details for
Quantity
  • Availability
  • Add to Cart