APOH316CDE11 Apollo Scientific Ltd

N-Benzyl-2-phenethylamine

  • MDL number MFCD00009624
  • Cas Number : 3647-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Density 1.007
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 162-165
Product Link http://www.apolloscientific.co.uk/display_item.php?id=130127

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=130127
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart