APOH316CDE12 Apollo Scientific Ltd

2-(6-Oxo-4-(trifluoromethyl)pyrimidin-1(6H)-yl)acetic acid

  • MDL number MFCD27974757
  • Cas Number : 1708178-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131315

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=131315
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart