APOH316CDE1B Apollo Scientific Ltd

4-Bromo-1-(4-chlorophenyl)-4,4-difluorobutane-1,3-dione

  • MDL number MFCD29042701
  • Cas Number : 1823188-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131299

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=131299
Availability details for
Quantity
  • Availability
  • Add to Cart