APOH316CDE20 Apollo Scientific Ltd

(E)-1-(2,6-Difluorophenyl)-3-(dimethylamino)prop-2-en-1-one

  • MDL number MFCD02089573
  • Cas Number : 138716-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131307

Safety Information

signalword Warning
pcodes P280;P301+P312;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=131307
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart