APOH316CDE24 Apollo Scientific Ltd

8-Chloro-3-methyl-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine

  • MDL number MFCD12406874
  • Cas Number : 1823182-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131274

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=131274
Availability details for
Quantity
  • Availability
  • Add to Cart