APOH316CDE2F Apollo Scientific Ltd

4-Chloro-2-(methylsulfonyl)-6-(trifluoromethyl)pyrimidine

  • MDL number MFCD31616619
  • Cas Number : 119610-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131264

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=131264
Availability details for
Quantity
  • Availability
  • Add to Cart