APOH316CDE30 Apollo Scientific Ltd

4-(Trifluoromethyl)-1H-imidazole-2(3H)-thione

  • MDL number MFCD28676943
  • Cas Number : 149432-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131266

Safety Information

signalword Warning
pcodes P280;P304+P340;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=131266
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart