APOH316CDE31 Apollo Scientific Ltd

2,2-Difluoro-N-(2-hydroxyethyl)-2-(pyridin-2-yl)acetamide

  • MDL number MFCD21337845
  • Cas Number : 1823183-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131271

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=131271
Availability details for
Quantity
  • Availability
  • Add to Cart