APOH316CDE35 Apollo Scientific Ltd

6-(Trifluoromethoxy)spiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride

  • MDL number MFCD29042696
  • Cas Number : 1823184-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-trifluoromethoxyspirobenzoe13oxazine-24-piperidin-43h-one-hydrochloride

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/6-trifluoromethoxyspirobenzoe13oxazine-24-piperidin-43h-one-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart