APOH316CDE36 Apollo Scientific Ltd

8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid

  • MDL number MFCD12406878
  • Cas Number : 1823188-42-6
  • Molecular Weight : 264.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Inchi Code 1S/C9H4ClF3N2O2/c10-5-1-4(9(11,12)13)3-15-6(8(16)17)2-14-7(5)15/h1-3H,(H,16,17)
Inchi Key XAXCHISMSFVBBE-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-chloro-6-trifluoromethylimidazo12-apyridine-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/8-chloro-6-trifluoromethylimidazo12-apyridine-3-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart