APOH316CDE39 Apollo Scientific Ltd

1-(4-Fluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone

  • MDL number MFCD03411212
  • Cas Number : 58905-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131288

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=131288
Availability details for
Quantity
  • Availability
  • Add to Cart